EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A simple method for the qualitative demonstration of large and small plasmid deoxyribonucleic acids in gram-positive and gram-negative bacteria.
PMID 6810724