EMAIL THIS PAGE TO A FRIEND
Genetics
Application of the ovarian teratoma mapping method in the mouse.
PMID 6852526
Abstract
Murine ovarian teratomas were used to determine recombination percentages for gene-gene and centromere-gene intervals. Data were obtained utilizing a recombinant inbred strain, LTXBJ, and a number of newly developed LT/SvEi congenic strains.--Centromere-gene recombination was measured at 11.3 +/- 1.2% for the centromere of chromosome 7 - Gpi-1 interval and 15.8 +/- 2.4% for the centromere of chromosome 14 - Np-1 interval using the ovarian teratoma method. The centromere - Np-1 interval was measured at 26.5 +/- 3.6% using a standard backcross involving the Rb6Bnr Robertsonian translocation as a centromere marker.--To assess the accuracy of the ovarian teratoma mapping method, we compared the recombination frequency obtained for the Mpi-1-Mod-1 interval on chromosome 9 using the ovarian teratoma method to that obtained using a standard backcross. The recombination percentage was 22.9 +/- 5.4 using the ovarian teratoma method and 18.6 +/- 3.3 using the backcross method, indicating that the two methods produce equivalent estimates of recombination. In addition, for centromere-gene intervals known to be more than 30 cM in length, the ovarian teratoma method was consistent with classical recombination methods, yielding high recombination percentages. We conclude from these results that the ovarian teratoma mapping method is a reliable method for estimating recombination frequencies and the most accurate method available for estimating centromere-gene recombination frequency in the mouse.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0505
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate
C21H27N7NaO17P3 · xH2O