EMAIL THIS PAGE TO A FRIEND
Clinical science (London, England : 1979)
3-Methylhistidine as a measure of skeletal muscle protein breakdown in human subjects: the case for its continued use.
PMID 6872455
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
67520
1-Methyl-L-histidine, ≥98.0% (TLC)
C7H11N3O2