EMAIL THIS PAGE TO A FRIEND
FEBS letters
Intracellular pH and free calcium changes in single cells using quene 1 and quin 2 probes and fluorescence microscopy.
PMID 6884525
Abstract
Photometric fluorescence microscopy has been used to measure intracellular pH (pHi) and free calcium concentrations [( Ca]i) in individual mouse thymocytes and 2H3 rat basophil leukaemic cells containing indicators for pH (quene 1) or calcium (quin 2). The pHi and [Ca]i measurements in individual 2H3 cells and mouse thymocytes and their responses to various stimuli were consistent with the corresponding data obtained from suspensions of these cells measured in a spectrofluorimeter. Photometric fluorescence microscopy of these indicators in individual cells provides a sensitive and fast method of following pHi and [Ca]i responses in individual cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q1626
Quene 1-AM
C38H41N3O17