Description
Molecular Formula
Add to Cart
G0513
Gly-Pro p-nitroanilide hydrochloride, ≥99% (HPLC)
C13H16N4O4 · HCl
G2901
Gly-Pro p-nitroanilide p-toluenesulfonate salt, ~99% (TLC)
C13H16N4O4 · C7H8O3S