EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
N-(alpha-Hydroxyalkanoyl) derivatives of Leu-Val-Phe-OCH3 as inhibitors of renin.
PMID 6993683
Abstract
The following N-(alpha-hydroxylakanoyl) derivatives of Leu-Val-Phe-OCH3 were synthesized and tested for their ability to inhibit human amniotic renin: D- and L-alpha-hydroxyisocaproyl-Leu-Val-Phe-OCH3, D- and L-alpha-hydroxyisovaleryl-Leu-Val-Phe-OCH3, L-2-hydroxy-3-phenylpropanoyl-Leu-Val-Phe-OCH3, and D- and L-alpha-hydroxyphenylacetyl-Leu-Val-Phe-OCH3. Analysis of the compounds through the use of Dixion plots showed all of the compounds to be competitive inhibitors of renin. All but D-alpha-hydroxyisovaleryl-Leu-Val-Phe-OCH3 were found to be more active than the known tetrapeptide inhibitor Leu-Leu-Val-Phe-OCH3 (1). The two most active compounds of the series were L-alpha-hydroxyisocaproyl-Leu-Val-Phe-OCH3 (Ki = 0.23 mM) and L-alpha-hydroxyisovaleryl-Leu-Val-Phe-OCH3 (Ki = 0.3 mM).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55452
D-α-Hydroxyisovaleric acid, ≥98.0% (T)
C5H10O3