EMAIL THIS PAGE TO A FRIEND
Vestnik rentgenologii i radiologii
[Tantalum dust as a contrast substance for laryngography and bronchography (a review of the literature)].
PMID 7013251
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262889
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Ta
357251
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Ta
262897
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Ta
357243
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Ta
262919
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Ta
545007
Tantalum, powder, 99.99% trace metals basis
Ta
692824
Tantalum, powder, 60-100 mesh, 99.9%
Ta
262846
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Ta
593486
Tantalum, nanopowder, <25 nm particle size (BET), ≥99% trace metals basis
Ta
262854
Tantalum, rod, diam. 6.35 mm, ≥99.9% trace metals basis
Ta
356980
Tantalum, rod, diam. 3.2 mm, ≥99.9% trace metals basis
Ta
262862
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Ta
357006
Tantalum, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ta
767514
Tantalum, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ta
GF34465888
Tantalum, foil, 0.5m coil, thickness 0.034mm, as rolled, 99.9%
Ta
GF96084603
Tantalum, foil, 0.5m coil, thickness 0.03mm, as rolled, 99.9%
Ta
GF78648092
Tantalum, foil, 0.5m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF38365106
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76540181
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta
GF08928108
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 1mm, annealed, 99.9%
Ta
GF61023574
Tantalum, foil, 0.5m coil, thickness 0.05mm, annealed, 99.9%
Ta
GF07210347
Tantalum, foil, 10m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF10417698
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53229269
Tantalum, foil, 10m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76534500
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Ta
GF80276182
Tantalum, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Ta
GF85784653
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Ta
GF65422739
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, flattened, 99.9%
Ta
GF86342485
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Ta
GF60298195
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.9%
Ta