EMAIL THIS PAGE TO A FRIEND
Klinicheskaia meditsina
[Clinical use of furazolidone].
PMID 7040784
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9505
Furazolidone
C8H7N3O5
46297
Furazolidone, VETRANAL, analytical standard
C8H7N3O5