Description
Molecular Formula
Add to Cart
791768
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
S7795
Sodium carbonate, BioXtra, ≥99.0%
CNa2O3
204420
Sodium carbonate, 99.995% trace metals basis
CNa2O3
451614
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
CNa2O3
223484
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
CNa2O3
222321
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
CNa2O3
223530
Sodium carbonate, powder, ≥99.5%, ACS reagent
CNa2O3
S2127
Sodium carbonate, ReagentPlus®, ≥99.5%
CNa2O3
71345
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
CNa2O3
31432
Sodium carbonate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance)
CNa2O3
13418
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
CNa2O3
NIST351A
Sodium carbonate, NIST® SRM® 351a
CNa2O3
71363
Sodium carbonate, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
CNa2O3
28-2170
Sodium carbonate, JIS special grade, ≥99.5%
CNa2O3
NIST2192A
Sodium carbonate, NIST SRM 2192a, pD Standard
CNa2O3
900503
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
900505
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
CNa2O3
900504
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
CNa2O3
1613757
Sodium carbonate anhydrous, United States Pharmacopeia (USP) Reference Standard
CNa2O3
S4132
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
CNa2O3 · H2O
13568
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · H2O
230952
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
CNa2O3 · H2O