EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Duplex regions in "single-stranded" phiX174 DNA are cleaved by a restriction endonuclease from Haemophilus aegyptius.
PMID 71298
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R5628 Hae III from Haemophilus aegyptius, Restriction Enzyme