EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Covalent binding of a neurotoxic n-hexane metabolite: conversion of primary amines to substituted pyrrole adducts by 2,5-hexanedione.
PMID 7157375