EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Separation of enantiomeric amines by ion-pair chromatography.
PMID 7217250
Abstract
A high-performance liquid chromatographic method for the separation of optical isomers (enantiomers) of amines is described. It is based on ion-pair chromatography with a chiral counter ion in a system with an organic mobile phase and an adsorbing stationary phase. The method has been applied to enantiomers of 1-aryloxy-3-isopropylamine-2-propanol derivatives (alprenolol, metoprolol, propranolol) which are completely resolved with (+)-10-camphorsulphonate as the counter ion. Studies of the influence of the counter-ion structure and the mobile phase composition are presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21360
(+)-Camphor-10-sulfonic acid, purum, ≥98.0% (T)
C10H16O4S