EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Convenient synthesis of alkyl esters of urocanic acid.
PMID 7229940
Abstract
Ethyl, n-dodecyl, and n-hexadecyl esters of urocanic acid (4-imidazoleacrylic acid) were prepared from 4-imidazolecarboxaldehyde in satisfactory yields via the Wittig reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
456128
4-Imidazolecarboxaldehyde, 98%
C4H4N2O