EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Phenyl 2-acetamido-2-deoxy-3-O-beta-D-galactopyranosyl-alpha-D-galactopyranoside, a substrate for sialyltransferase.
PMID 7237483
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5019
Benzyl 2-acetamido-2-deoxy-3-O-β-D-galactopyranosyl-α-D-galactopyranoside
C21H31NO11