EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Purification, enzymatic properties, and active site environment of a novel manganese(III)-containing acid phosphatase.
PMID 7287728
Abstract
A new manganese-containing acid phosphatase has been isolated and crystallized from sweet potato tubers. The pure enzyme contains one atom of manganese per Mr = 110,000 polypeptide and shows phosphatase activity toward various phosphate substrates. The pH optimum of the enzyme was 5.8 and the enzyme activity was inhibited by Cu2+, Zn2+, Hg2+, AsO43-, and MoO42-. This stable metalloenzyme is red-violet in color with an intense absorption band at 515 nm (epsilon - 2460). Our electronic, circular dichroism, and electron spin resonance findings strongly indicate that the Mn-valence state of the native enzyme is trivalent. When the Mn-enzyme is excited by the 5145 A line of Ar+ laser, prominent Raman lines at 1230, 1298, 1508, and 1620 cm-1 were detected. This Raman spectrum can probably be interpreted in terms of internal vibration of a coordinated tyrosine phenolate anion. The tryptophan-modified enzyme showed a positive Raman band at 370 cm-1, which is preferentially assigned to a Mn(III)-S streching mode. The modification of the Mn-enzyme by N-bromosuccinimide led to a large decrease in the fluorescence intensity of 335 nm which was dominated by its tryptophan residues within a considerable hydrophobic environment. The acid phosphatase activity was significantly decreased by the tryptophan modification. With respect to the active site donor sets, the Mn(III)-containing acid phosphatase is distinctly different from the Zn(II)-containing alkaline phosphatase. Of interest is also the appreciable similarity of some enzymatic and spectroscopic properties between the present enzyme and uteroferrin.