EMAIL THIS PAGE TO A FRIEND
Journal of steroid biochemistry
Metabolism of 1,4-androstadiene-3,17-dione by human placental microsomes. Enzyme properties and kinetic parameters in the formation of estrogens and 17 beta-hydroxy-1,4-androstadien-3-one.
PMID 7311525
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7505
1,4-Androstadiene-3,17-dione
C19H24O2