EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
The rapid, quantitative determination of neutral sugars (as aldononitrile acetates) and amino sugars (as O-methyloxime acetates) in glycoproteins by gas--liquid chromatography.
PMID 7356120