EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
A new endogenous opioid peptide from bovine adrenal medulla: isolation and amino acid sequence of a dodecapeptide (BAM-12P).
PMID 7417331