EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Increased ability of ethylnitrosourea-exposed brain cells to survive suspension in agar.
PMID 7426322
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93040125
BE10-7