EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
An improved spectrophotometric assay for human plasma carboxypeptidase N1.
PMID 7457880
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5882
N-(3-[2-Furyl]acryloyl)-Ala-Lys, powder
C16H23N3O5