EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Vitamin K-dependent carboxylase: purification of the rat liver microsomal enzyme.
PMID 7458333
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5523
Phe-Leu-Glu-Glu-Leu, ≥97% (HPLC)
C31H47N5O10