EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Neuroprotective properties of the uncompetitive NMDA receptor antagonist remacemide hydrochloride.
PMID 7486610
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0335
Remacemide hydrochloride, ≥98% (HPLC)
C17H20N2O · HCl