EMAIL THIS PAGE TO A FRIEND
Archives des maladies du coeur et des vaisseaux
[Reflections of an expert committee of the French Society of Cardiology on the use of methylergometrine maleate (Methergin) for the detection of abnormal coronary vasomotricity].
PMID 7487274
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1430000
Methylergonovine maleate, United States Pharmacopeia (USP) Reference Standard
C20H25N3O2 · C4H4O4