EMAIL THIS PAGE TO A FRIEND
Stroke
The pharmacology of AMPA receptors and their antagonists.
PMID 7504337
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0326
(S)-AMPA, ≥98%
C7H10N2O4