EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Formation of 8-nitroguanine by the reaction of guanine with peroxynitrite in vitro.
PMID 7554052
Abstract
Nitric oxide and superoxide anion, both formed in inflamed tissues, react rapidly to form the peroxynitrite anion (ONOO-), a strong oxidant which can initiate reactions characteristic of hydroxyl radical (HO.), nitronium ion (NO2+) and nitrogen dioxide radical (NO2.). Peroxynitrite, therefore, may cause DNA or tissue damage, contributing to the multistage carcinogenesis process. We have studied reactions of various bases, nucleosides or deoxynucleosides with peroxynitrite in vitro. Guanine reacted rapidly with peroxynitrite under physiological conditions and formed several substances, two of which were yellow, a characteristic of nitro and nitroso compounds. On the basis of chromatographic and spectral evidence we identified the major compound (which accounts for approximately 80% of all compounds formed) as 8-nitroguanine. Its formation was maximal at approximately pH 8 and increased dose-dependently with peroxynitrite concentration, but was not dependent on guanine concentration. The presence of ferric ions, which has been shown to catalyse nitration of tyrosine, did not affect nitration of guanine. 8-Nitroguanine could act as a specific marker for DNA damage induced by peroxynitrite in inflamed tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G11950
Guanine, 98%
C5H5N5O