EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
An unstable small-colony variant of a noninvasive mutant of Salmonella typhimurium is highly invasive for MDCK cells.
PMID 7557295
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
84121903
MDCK Cell Line canine