EMAIL THIS PAGE TO A FRIEND
Bioconjugate chemistry
A convenient route to thiol terminated peptides for conjugation and surface functionalization strategies.
PMID 7632807
Abstract
The derivatization of poly(p-(chloromethyl)styrene-co-divinylbenzene) (Merrifield resin) with N-(tert-butoxycarbonyl)-2-aminoethanethiol is presented as a convenient route for the generation of thiol terminated peptides using a solid phase methodology. Maximum resin substitution reached 92% (773 mumol/g) after 24 h. However, at 30 min, yields exceeded 400 mumol/g, above which the resin is suitable for solid phase peptide synthesis. Thiol terminated peptides are well-suited for subsequent chemical conjugation reactions or for the formation of organic monolayers on metal substrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
458910
2-(Boc-amino)ethanethiol, 97%
C7H15NO2S
17353
di-Boc-cystamine, ≥98.0% (TLC)
C14H28N2O4S2