EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
mSlo, a complex mouse gene encoding "maxi" calcium-activated potassium channels.
PMID 7687074
Abstract
Complementary DNAs (cDNAs) from mSlo, a gene encoding calcium-activated potassium channels, were isolated from mouse brain and skeletal muscle, sequenced, and expressed in Xenopus oocytes. The mSlo-encoded channel resembled "maxi" or BK (high conductance) channel types; single channel conductance was 272 picosiemens with symmetrical potassium concentrations. Whole cell and single channel currents were blocked by charybdotoxin, iberiotoxin, and tetraethylammonium ion. A large number of variant mSlo cDNAs were isolated, indicating that several diverse mammalian BK channel types are produced by a single gene.