EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Comparison of FK-506, rapamycin, ascomycin, and cyclosporine in mouse models of host-versus-graft disease and heterotopic heart transplantation.
PMID 7689812
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3835
Ascomycin from Streptomyces hygroscopicus var. ascomyceticus
C43H69NO12