EMAIL THIS PAGE TO A FRIEND
The American journal of cardiology
Efficacy and safety of nisoldipine coat core in the management of angina pectoris, systemic hypertension, and ischemic ventricular dysfunction.
PMID 7726125
Abstract
The effects of the long-acting dihydropyridine calcium antagonist nisoldipine coat core (CC) have been investigated in > 3,500 patients with angina pectoris, hypertension, and ischemic ventricular dysfunction. In patients with angina pectoris, nisoldipine CC improved total treadmill exercise duration (p = 0.027), delayed the onset of angina pectoris (p = 0.009), and increased time to exercise-induced ST-segment depression (p = 0.061). In general, nisoldipine 20-40 mg was effective, and the dose-response curve flattened thereafter. In patients with hypertension, 10-40 mg once daily as monotherapy reduced blood pressure (p < 0.05), with a fall in diastolic pressure of > or = 10 mm Hg or a final diastolic pressure of < 90 mm Hg in 35-63% of patients. In most patients followed for a year, nisoldipine CC was continued as monotherapy. Efficacy was similar in patients < 65 and > 65 years of age. In the Doppler Flow and Echocardiography in Functional Cardiac Insufficiency: Assessment of Nisoldipine Therapy (DEFIANT-I) study of patients recovering from myocardial infarction, nisoldipine CC had a salutary effect on diastolic ventricular function, with a higher transmitral early filling velocity and shorter isovolumic relaxation time than in patients receiving placebo. Bicycle exercise capacity was greater (by 12 W; 95% confidence interval, 0.8-23.3) and exercise-induced ischemia occurred less frequently. The nisoldipine CC data pool (3,679 patients) showed that the drug was well tolerated with a low incidence of side effects.(ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0165
Nisoldipine, ≥98% (HPLC)
C20H24N2O6