EMAIL THIS PAGE TO A FRIEND
Bulletin du cancer
The pharmacologic modulation of 5-fluorouracil with folinic acid, methotrexate, trimetrexate, and n-phosphonacetyl-l-aspartic acid (PALA). Mechanisms of the interactions and clinical data.
PMID 7727863