EMAIL THIS PAGE TO A FRIEND
Current opinion in lipidology
Apolipoprotein B messenger RNA editing: insights into the molecular regulation of post-transcriptional cytidine deamination.
PMID 7773570
Abstract
A site-specific cytidine deamination (cytidine to uridine) in nuclear apolipoprotein B messenger RNA creates a translational stop codon that produces apolipoprotein B48. This process is mediated by an enzyme composed of distinct subunits, including apolipoprotein B messenger RNA editing enzyme catalytic polypeptide-1 and additional complementation factors. The apolipoprotein B messenger RNA editing enzyme catalytic polypeptide-1 is expressed ubiquitously in the rat, but is largely confined to the small intestine in humans and rabbits. By contrast, complementation activity is present in tissues that neither express nor edit apolipoprotein B messenger RNA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30270
Cytidine, ≥99.0% (HPLC)
C9H13N3O5