EMAIL THIS PAGE TO A FRIEND
Drugs
Isradipine. An update of its pharmacodynamic and pharmacokinetic properties and therapeutic efficacy in the treatment of mild to moderate hypertension.
PMID 7789292
Abstract
Since the earlier review in Drugs substantial additional data have accumulated regarding the antihypertensive efficacy of isradipine in various clinical situations, as well as data on its clinical effects in atherosclerosis. Recent therapeutic trials confirm that the efficacy of isradipine in the treatment of patients with mainly mild to moderate hypertension, when administered orally as a conventional or modified release preparation, is similar to that of titrated dosages of amlodipine, felodipine, nifedipine, diltiazem, captopril, methyldopa, metoprolol, prazosin and hydrochlorothiazide. A further decrease in blood pressure can be expected when isradipine is combined with another antihypertensive drug in patients who have not responded adequately to monotherapy. Initial studies have shown that intravenous isradipine is effective in controlling hypertension following coronary artery bypass graft surgery and that it appears useful in the treatment of intraoperative hypertension and hypertensive crisis, and in hypertensive disorders in pregnancy, when administered orally or intravenously. A large study, the Multicentre Isradipine Diuretic Atherosclerosis Study (MIDAS), was designed to compare the efficacy of isradipine and hydrochlorothiazide in reducing the rate of progression of carotid artery wall thickness, measured by B-mode ultrasound, as a surrogate for early atherosclerosis. Results indicated that wall thickness increased significantly less with isradipine than hydrochlorothiazide after 6 months of therapy. Thereafter the rate of progression remained parallel for the remainder of the 3-year trial. The confirmation of its antihypertensive efficacy, along with its favourable haemodynamic profile and reversal of left ventricular hypertrophy, minimal effect on glucose and lipid metabolism, preservation of quality of life and good tolerability, makes isradipine a suitable drug for the treatment of most patients with mild to moderate hypertension.
Related Materials