EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Flavonoid antioxidants: rate constants for reactions with oxygen radicals.
PMID 7808314
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9283
Luteolin, ≥98% (TLC), powder
C15H10O6