EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Up-regulation of [3H]-des-Arg10-kallidin binding to the bradykinin B1 receptor by interleukin-1 beta in isolated smooth muscle cells: correlation with B1 agonist-induced PGI2 production.
PMID 7834189
Abstract
1. Binding of the specific bradykinin B1 receptor agonist, [3H]-des-Arg10-kallidin (-KD) was investigated in smooth muscle cells (SMC) isolated from rabbit mesenteric arteries (RMA). 2. [3H]-des-Arg10-KD specifically bound to interleukin-1 (IL-1)-treated RMA-SMC in a saturable fashion with an equilibrium dissociation constant (KD) of 0.3-0.5 nM. The number of binding sites per cell was 20,000-35,000. Kinins inhibited [3H]-des-Arg10-KD binding to RMA-SMC with an order of potency very similar to that observed in typical B1 specific bioassays: des-Arg9-bradykinin (BK) approximately KD > BK. Furthermore, the B1 receptor antagonist [Leu8]des-Arg9-BK inhibited [3H]-des-Arg10-KD binding with an IC50 of 43 nM as expected for its effect at B1 receptors. The B2 receptor antagonists, NPC 567 and Hoe 140 only affected [3H]-des-Arg10-KD binding at very high concentrations (IC50 = 0.8 microM and IC50 > 10 microM, respectively). 3. Des-Arg9-BK (B1 agonist) and [Hyp3]Tyr(Me)8-BK (B2 agonist) did not induce prostacyclin (PGI2) production by RMA-SMC. Lipopolysaccharide (LPS) treatment of the cells did not affect the B1 agonist response whereas IL-1 beta treatment produced a 7 fold increase in des-Arg9-BK-stimulated PGI2 production. IL-1 beta also stimulated the response to B2 agonists. 4. Des-Arg9-BK-induced PGI2 secretion in IL-1-primed RMA-SMC was mediated by B1 receptors since it was inhibited by [Leu8]des-Arg9-BK (IC50 = 56-73 nM) but not by Hoe 140. High concentrations of NPC 567 (IC5o = 2.4 micro M) were required to inhibit PGI2 production induced by B1 agonists.5. IL- 1-treated RMA-SMC displayed a 5 fold increase in the number of B1 receptors without modification of the affinity constant, thus establishing a possible relationship between the receptor density and the IL-i-primed B1 response.6. LPS treatment of the cells induced a 4 fold increase in B1 receptor number without modifying PGI2 secretion. This observation suggests that IL-1 but not LPS, in addition to increase in the number of receptors, signals the cell to permit the coupling of B1 receptors to the PLA2/cyclo-oxygenase pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1542
[Lys-des-Arg9]-Bradykinin, ≥95% (HPLC)
C50H73N13O11