EMAIL THIS PAGE TO A FRIEND
Trends in pharmacological sciences
GABAB receptor antagonists: from synthesis to therapeutic applications.
PMID 7905214
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5851
CGP 35348 hydrate, ≥97% (NMR), solid
C8H20NO4P · xH2O