EMAIL THIS PAGE TO A FRIEND
The Japanese journal of antibiotics
[Reevaluation of current antimicrobials. Cefaclor].
PMID 7933517
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72579
Cefaclor, analytical reference material
C15H14ClN3O4S
C6895
Cefaclor
C15H14ClN3O4S