EMAIL THIS PAGE TO A FRIEND
Pharmacological reviews
International Union of Pharmacology classification of prostanoid receptors: properties, distribution, and structure of the receptors and their subtypes.
PMID 7938166
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5640
Prostaglandin E2, ≥93% (HPLC), synthetic
C20H32O5