EMAIL THIS PAGE TO A FRIEND
Nutrition reviews
Mechanism of the mitogenic and carcinogenic action of fumonisin B1, a mycotoxin.
PMID 8090377
Abstract
Fumonisins, mycotoxins found on corn and other grains, can act as carcinogens. Recent studies have shown that fumonisin B1 stimulates cell proliferation due to inhibition of sphinganine N-acyl-transferase, an important rate limiting enzymatic step in sphingolipid biosynthesis. Subsequent accumulation of free sphingoid bases in the cell can act as tumor promoters.
Related Materials