EMAIL THIS PAGE TO A FRIEND
Otolaryngologia polska = The Polish otolaryngology
[Eight-year long study of the usage of flunisolide (syntaris) at the Otolaryngological clinic at the Warsaw Academy].
PMID 8090503
Abstract
Brief information about clinical experience on flunisolide in nasal pathology treatment is presented in this paper. The authors own observations are also included. They describe some cases histories as to how flunisolide can act positively and negatively at on nasal and paranasal clinical pathology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5021
Flunisolide, ≥97%
C24H31FO6