EMAIL THIS PAGE TO A FRIEND
Acta biochimica Polonica
The control of L-phenylalanine ammonia-lyase activity by phosphonate and aminooxy analogues of phenylalanine.
PMID 8140817
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1530503
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
C9H11NO2