EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Comparison of properties of agarose for electrophoresis of DNA.
PMID 8227266
Abstract
Agarose as a medium for separation of DNA was first introduced in 1962 and since the early 1970s agarose submarine gel electrophoresis has been synonymous with separations of DNA molecules larger than 1 kilobase pair (kb). The large pore size, low electroendosmosis and strength of the matrix have advantages over other media such as polyacrylamide for many applications. The variety of grades of agarose, developed by chemical manipulation of the substitutions on the agarose polymer, provides a range of matrices for separation of DNA molecules from a few base pairs (bp) to over 5 megabase pairs (Mb) in length. The introduction of low-melting-temperature agarose has revolutionised the extraction and manipulation of chromosome-sized molecules. On the other hand, the demand for analysis of very small quantities of DNA will most likely lead to the increasing importance of capillary electrophoresis. Many theories have been propounded to explain the electrophoretic migration of DNA in agarose. The most popular of these has been reptation theory but none can account for all of the reported anomalies in migration. However, anomalous migration has been exploited to study DNA structure, topology and catenation. An example of the use of two-dimensional electrophoresis to demonstrate the complexity of DNA migration through agarose is given. Generally, for molecules smaller than 50 kb, electrophoretic separation is a function of length. By alternately electrophoresing DNA in two different directions, molecules as large as 5.7 Mb have been effectively separated, although with such large molecules DNA structure as well as size may determine migration. In the case of separations of chromosomes from the intestinal protozoan, Giardia duodenalis, for example, a discrepancy of 1 Mb in the size of one chromosome, with an apparent size of 0.7-2.0 Mb, depended on the boundary conditions of separation. Major challenges for the molecular biologist are separation of larger chromosomal sized molecules, greater number of samples and smaller formats. Towards this challenge computer-aided technology is a key component in the control of electrophoresis parameters and analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9539
Agarose, BioReagent, for molecular biology, low EEO
A6013
Agarose, Type I, low EEO
A6877
Agarose, Medium EEO, for molecular biology
A0169
Agarose, Type I-A, low EEO
A4718
Agarose, High resolution, for molecular biology
A4679
Agarose, Low EEO, for Immunoelectrophoresis
A7431
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
A9918
Agarose, Type II-A, Medium EEO
A0576
Agarose, Low EEO
A2790
Agarose, Wide range, for molecular biology
A9311
Agarose, Medium EEO, for electrophoresis
A3893
Agarose, Type VI, High Gelling Temperature
A2929
Agarose, For pulsed field electrophoresis running gel
A3038
Agarose, Type XI, low gelling temperature, suitable for separation of small nucleic acid fragments
A5030
Agarose, Ultra-low Gelling Temperature
A3643
Agarose, Type IV, Special High EEO
A2576
Agarose, Ultra-low Gelling Temperature, molecular biology grade
A9793
Agarose, High EEO
A4905
Agarose, Type VIII
A7174
Agarose, High Gelling Temperature
A5304
Agarose, Special High EEO
A3054
Agarose, For pulsed field electrophoresis sample preparation
05066
Agarose, High EEO, for molecular biology
4B200
Sepharose® 4B, 45-165 μm bead diameter
GE17-0120-01
Sepharose® 4B, GE Healthcare, 17-0120-01, pack of 1 L
6B100
Sepharose® 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
GE17-0110-01
Sepharose® 6B, GE Healthcare, 17-0110-01, pack of 1 L
U0507
Ultrogel® A4, aqueous ethanol suspension, 60-140 μm (wet bead diameter), exclusion limit 20,000,000 Da, fractionation range 55,000-9,000,000 Da (globular proteins)