EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Is the "preamyloid" of diffuse plaques in Alzheimer's disease really nonfibrillar?
PMID 8256851
Abstract
Several papers have described an 'amorphous' component of the amyloid in diffuse plaques and it has been suggested that this is 'preamyloid,' which is not organized into fibrils. Because most of the studies have been performed on autopsy tissue it was the purpose of this study to compare the ultrastructure of diffuse amyloid deposits in well preserved Alzheimer's disease biopsy specimens with autopsy tissues from patients with Alzheimer's disease and Down's syndrome. A postembedding immunogold technique with anti-beta/A4 protein demonstrated gold particles exclusively on extracellular amyloid fibrils in both biopsy and autopsy brains. We have presented evidence that suggests the claim for the existence of an amorphous component within the beta/A4 protein-positive material is unconvincing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6402 Anti-τ (Tau) antibody produced in rabbit, whole antiserum