EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Starch gel electrophoresis in vertebrates.
PMID 8264419
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5651
Starch from potato, for electrophoresis