EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Effects of transforming growth factor beta and interleukin-1 beta on expression of cyclooxygenase 1 and 2 and phospholipase A2 mRNA in lung fibroblasts and endothelial cells in culture.
PMID 8280164
Abstract
Experiments were conducted to determine the roles of the rate limiting enzymes, cyclooxygenase 1 and 2 (COX1 and COX2) and cytoplasmic phospholipase A2 (PLA2), in transforming growth factor beta (TGF-beta) and interleukin-1 (IL-1 beta) activated prostaglandin synthesis. Results show that TGF-beta increases steady state levels of COX1 mRNA in both human embryo lung fibroblasts (IMR90) and calf pulmonary artery endothelial cells (BPAEC). Temporal experiments show that TGF-beta increases, within 2hrs, a 5.5kb COX1 in IMR90 and the 2.7kb COX1 mRNA in BPAEC. IL-1 beta increases COX1 mRNA only in IMR-90, not BPAEC. COX2 mRNA, under basal conditions, is not detected in BPAEC and is expressed only marginally in IMR90. TGF-beta or IL-1 beta have no effect on expression of COX2 gene in either cell type. IL-1 beta increases steady state levels of PLA2 mRNA in both IMR90 and BPAEC while TGF-beta increases expression of the PLA2 gene only in BPAEC. Time experiments with TGF-beta show induction of PLA2 mRNA within 1hr, peaking at 4hrs. PG synthesis in response to the cytokines was determined in IMR90 and BPAEC to further assess the significance of the above results. TGF-beta increases the synthesis of prostacyclin in BPAEC in a time related fashion peaking at 8hrs at 13 fold above basal. To focus on the action of COX1 and bypass the action of PLA2, exogenous arachidonic acid was used as substrate for PG synthesis. In these experiments IL-1 beta increases PGE2 synthesis 8 fold in IMR90 while IL-1 beta and TGF-beta added simultaneously increases PGE2 synthesis 25 fold. These results in sum illustrate that the cytokines, TGF-beta and IL-1 beta, regulate both COX1 and PLA2 mRNA levels. Furthermore, this regulation appears coordinated to bring about elevation of prostaglandin synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86123101
BPAEC