EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
3-substituted thieno[2,3-b][1,4]thiazine-6-sulfonamides. A novel class of topically active carbonic anhydrase inhibitors.
PMID 8295211
Abstract
3-Aminoalkyl derivatives of thieno[2,3-b][1,4]thiazine-6-sulfonamide were prepared for evaluation as topically active ocular hypotensive agents. The compounds described were found to be excellent in vitro inhibitors of carbonic anhydrase II and in vivo to lower intraocular pressure in three rabbit models of ocular hypertension. Compounds 20A, 20B, and 20C met the requirement of formulation as a 1% solution at pH 5.2, but none of the compounds described exhibited greater activity in the normotensive albino rabbit, the alpha-chymotrypsin-treated albino rabbit, or the normotensive pigmented rabbit than MK-927 or MK-507, the present clinical candidates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
544272
5-Chlorothiophene-2-sulfonyl chloride, 96%
C4H2Cl2O2S2