EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
L-694,247: a potent 5-HT1D receptor agonist.
PMID 8298808
Abstract
1. The 5-hydroxytryptamine (5-HT) receptor binding selectivity profile of a novel, potent 5-HT1D receptor agonist, L-694,247 (2-[5-[3-(4-methylsulphonylamino)benzyl-1,2,4-oxadiazol-5-yl ]- 1H-indole-3-yl]ethylamine) was assessed and compared with that of the 5-HT1-like receptor agonist, sumatriptan. 2. L-694,247 had an affinity (pIC50) of 10.03 at the 5-HT1D binding site and 9.08 at the 5-HT1B binding site (sumatriptan: pIC50 values 8.22 and 5.94 respectively). L-694,247 retained good selectivity with respect to the 5-HT1A binding site (pIC50 = 8.64), the 5-HT1C binding site (6.42), the 5-HT2 binding site (6.50) and the 5-HT1E binding site (5.66). The pIC50 values for sumatriptan at these radioligand binding sites were 6.14, 5.0, < 5.0 and 5.64 respectively. Both L-694,247 and sumatriptan were essentially inactive at the 5-HT3 recognition site. 3. L-694,247, like sumatriptan, displayed a similar efficacy to 5-HT in inhibiting forskolin-stimulated adenylyl cyclase in guinea-pig substantia nigra although L-694,247 (pEC50 = 9.1) was more potent than sumatriptan (6.2) in this 5-HT1D receptor mediated functional response. L-694,247 (pEC50 = 9.4) was also more potent than sumatriptan (6.5) in a second 5-HT1D receptor mediated functional response, the inhibition of K(+)-evoked [3H]-5-HT release from guinea-pig frontal cortex slices. 4. The excellent agreement observed for L-694,247 between the 5-HTlD radioligand binding affinity and the functional potency confirm that the two functional models (the inhibition of forskolin-stimulated adenylyl cyclase in guinea-pig substantia nigra and the inhibition of K+-evoked [3H]-5-HT release from guinea-pig frontal cortex) do indeed reflect 5-HTID-mediated events.5. L-694,247 is a novel, highly potent 5-HTID/5-HTIB receptor ligand which should prove useful for the exploration of the physiological role of these receptors in animals.