EMAIL THIS PAGE TO A FRIEND
The Veterinary clinics of North America. Equine practice
Furosemide, bumetanide, and ethacrynic acid.
PMID 8299013
Abstract
Furosemide and, less commonly, bumetanide and ethacrynic acid are potent diuretics administered to horses for a variety of reasons, including prophylaxis of exercise-induced pulmonary hemorrhage. These drugs affect urine volume and composition, and furosemide has marked effects on plasma volume and composition and on systemic hemodynamics at rest and during exercise.