EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
Studies on the effect of mevinolin (lovastatin) and mevastatin (compactin) on the fusion of L6 myoblasts.
PMID 8302293
Abstract
The effect of mevastatin and mevinolin on the fusion of L6 myoblasts was studied. Both compounds were present inhibitors of myoblast fusion at concentrations as low as 0.25 microM, but fusion was restored when the inhibitors were removed. Both compounds resulted in decreased binding of conA and WGA to cell surface oligosaccharides showing they were causing a reduction in N-linked cell surface glycoproteins. There was a reduction in creatine phosphokinase activities in the presence of both compounds showing that they were affecting biochemical differentiation. The presence of both compounds inhibited the incorporation of labeled mannose from GDP-mannose into lipid-sugar and N-linked glycoprotein, but the inhibition was reversed by addition of exogenous dolichol phosphate to the incorporation mixture. The main conclusion from these studies is that mevinolin and mevastatin are inhibiting myoblast fusion by affecting the synthesis of fusogenic cell surface N-linked glycoproteins probably by affecting the synthesis of dolichol phosphate containing oligosaccharides that are required as intermediates in N-linked glycoprotein biosynthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2537
Mevastatin, ≥95% (HPLC), powder
C23H34O5