EMAIL THIS PAGE TO A FRIEND
FEBS letters
Elongation factor-2 kinase: effective inhibition by the novel protein kinase inhibitor rottlerin and relative insensitivity towards staurosporine.
PMID 8307162
Abstract
The elongation factor-2 (eEF-2) is selectively phosphorylated by the eEF-2 kinase (calmodulin-dependent kinase III). This phosphorylation can be inhibited by calmodulin antagonists, such as CGS 9343B (IC50 = 4 microM). The novel protein kinase inhibitor rottlerin is shown to suppress eEF-2 phosphorylation with an IC50 of 5.3 microM. By contrast, the eEF-2 kinase is rather resistant towards the potent but non-selective protein kinase inhibitor staurosporine (IC50 > 50 microM) and thus can be differentiated from most other protein kinases that are suppressed by staurosporine in the nM range.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R5648
Rottlerin
C30H28O8